missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Antibody (40F11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-10068-0.05mg
This item is not returnable.
View return policy
Description
Fibrinogen Monoclonal specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Immunoassay, Sandwich ELISA.
Specifications
| Fibrinogen | |
| Monoclonal | |
| Unconjugated | |
| Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Fibrinogen, Fibrin degradation products. | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Western Blot | |
| 40F11 | |
| Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
| FGA | |
| Fibrin degradation products. | |
| 0.05 mg | |
| Cell Cycle and Replication | |
| 2243 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction