missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Antibody (1F3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10066AF750
368.90 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Fibrinogen Monoclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Immunoassay, ELISA
Specifications
| Fibrinogen | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoassay, ELISA | |
| 1F3 | |
| Western Blot, Immunoassay, Sandwich ELISA | |
| Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Fibrin degradation products. | |
| 0.2 mL | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 2243 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction