missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Antibody (1F3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
£464.00
Specifications
| Antigen | Fibrinogen |
|---|---|
| Clone | 1F3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 350 |
Description
Fibrinogen Monoclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Immunoassay, ELISASpecifications
| Fibrinogen | |
| Western Blot | |
| Alexa Fluor 350 | |
| Mouse | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 50mM Sodium Borate | |
| 2243 | |
| IgG2b | |
| Protein A purified |
| 1F3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Fibrin degradation products. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title