missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrinogen alpha chain Recombinant Rabbit Monoclonal Antibody (7K8J1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537943
This item is not returnable.
View return policy
Description
Positive Samples: Human plasma, Rat liver Immunogen sequence: NDEGEGEFWL GNDYLHLLTQ RGSVLRVELE DWAGNEAYAE YHFRVGSEAE GYALQVSSYE GTAGDALIEG SVEEGAEYTS HNNMQFSTFD RDADQWEENC A.
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
Specifications
| Fibrinogen alpha chain | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06399 | |
| Fga | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 7K8J1 | |
| Unconjugated | |
| Fga | |
| Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain; Fibrinopeptide A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361969 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction