missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen alpha chain Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20645-1-AP-20UL
This item is not returnable.
View return policy
Description
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.Specifications
| Fibrinogen alpha chain | |
| Polyclonal | |
| Unconjugated | |
| FGA | |
| FGA, Fib2, fibrinogen alpha chain | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14161, 2243 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| E9PV24, P02671 | |
| Fga | |
| Fibrinogen alpha chain Fusion Protein Ag9567 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction