missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrinogen alpha chain Monoclonal Antibody (471CT12.1.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537571
This item is not returnable.
View return policy
Description
Fibrinogen alpha chain Monoclonal Antibody for Western Blot
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
Specifications
| Fibrinogen alpha chain | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| E9PV24, P02671 | |
| Fga | |
| KLH conjugated synthetic peptide between 420-448 amino acids from the N-terminal region of human FGA. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgM |
| Western Blot | |
| 471CT12.1.2 | |
| Unconjugated | |
| Fga | |
| Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen a chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogena chain; Fibrinopeptide A | |
| Mouse | |
| Euglobulin precipitation | |
| RUO | |
| 14161, 2243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction