missing translation for 'onlineSavingsMsg'
Learn More

Fibrin beta-chain Antibody (8E5), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18416378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416378 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416378 Supplier Novus Biologicals Supplier No. NBP281111MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Fibrin beta-chain Monoclonal antibody specifically detects Fibrin beta-chain in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrin beta-chain
Applications Flow Cytometry
Classification Monoclonal
Clone 800000
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p
Host Species Rabbit
Immunogen 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2244
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.