missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fibrin beta-chain Monoclonal specifically detects Fibrin beta-chain in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
Specifications
| Antigen | Fibrin beta-chain |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 800000 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p |
| Gene Symbols | FGB |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?