missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrillin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£259.00 - £451.00
Specifications
| Antigen | Fibrillin 2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen Reported in scientific literature (PMID 26679090). |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443321
|
Novus Biologicals
NBP1-88169-25ul |
25 μL |
£259.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750034
|
Novus Biologicals
NBP1-88169 |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Fibrillin 2 Polyclonal specifically detects Fibrillin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Fibrillin 2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH) | |
| Unconjugated | |
| RUO | |
| Human | |
| CCA, congenital contractural arachnodactyly, DA9, fibrillin 2, fibrillin 5, fibrillin-2 | |
| FBN2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen Reported in scientific literature (PMID 26679090). | |
| Polyclonal | |
| Rabbit | |
| Cytoskeleton Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2201 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GMCFSGLVNGRCAQELPGRMTKMQCCCEPGRCWGIGTIPEACPVRGSEEYRRLCMDGLPMGGIPGSAGSRPGGTGGNGFAPSGNGNGYGPGGTGFIPIPGGNGFSPGVGGAGVGAGGQGPIITGLTILNQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title