missing translation for 'onlineSavingsMsg'
Learn More

Fibrillin 2 Antibody (1C2), Novus Biologicals™

Product Code. 18338227 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338227 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338227 Supplier Novus Biologicals Supplier No. H00002201M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibrillin 2 Monoclonal antibody specifically detects Fibrillin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillin 2
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 1C2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias CCA, congenital contractural arachnodactyly, DA9, fibrillin 2, fibrillin 5, fibrillin-2
Host Species Mouse
Immunogen FBN2 (NP_001990.2, 2776 a.a. ~ 2876 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RQKRSIHEPDPTAVEQISLESVDMDSPVNMKFNLSHLGSKEHILELRPAIQPLNNHIRYVISQGNDDSVFRIHQRNGLSYLHTAKKKLMPGTYTLEITSIP
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 2201
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.