Learn More
Fibrillin 1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-26935-100UL
Description
The Fibrillin 1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Fibrillin 1 . This antibody recognizes human antigen. The Fibrillin 1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Fibrillin 1 | |
| 1000 μg/mL | |
| FBN, FBN1, fibrillin 1, MASS, MFS1, OCTD, SGS, WMS | |
| 100 μL | |
| 2200 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |