missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrillin 1 Monoclonal Antibody (CL0226)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534973
This item is not returnable.
View return policy
Description
Immunogen sequence: DVRPGYCYTA LTNGRCSNQL PQSITKMQCC CDAGRCWSPG VTVAPEMCPI RATEDFNKLC SVPMVIPGRP EYPPPPLGPI PPVLPVPPGF PPGPQIPVPR PPVEYLYPSR EPPRVLPVNV TDYCQLVRYL CQNGRCIPTP GSCRCEC Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.
Fibrillin 1 is an extracellular acidic protein with a high cysteine content and an extended thread-like shape with mosaic composition of different types of extracellular modules. Most of the fibrillin molecule is contributed by 47 epidermal growth factor-like (EGF-like) repeats; 43 of them have a consensus sequence for calcium binding (cb). Other modules including 8-cysteine motifs (seven), hybrid motifs (two), and unique domains (three) are interspersed throughout the molecule[3-7]. Fibrillin 1 contains a proline-rich region close to the N-terminus that is replaced by a glycine-rich region in Fibrillin 2. The fibrillins are found throughout the connective tissue as integral components of extended fibrils.
Specifications
| Fibrillin 1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35555 | |
| FBN1 | |
| Recombinant protein fragment of Human FBN1 (Product #RP-90822) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| CL0226 | |
| Unconjugated | |
| FBN1 | |
| ACMICD; AI536462; Asprosin; B430209H23; ECTOL1; FBN; Fbn1; Fbn-1; Fib-1; fibrillin 1; fibrillin 15; fibrillin-1; GPHYSD2; MASS; MFLS; MFS1; MP340; OCTD; putative; SGS; SSKS; tight skin; Tsk; WMS; WMS2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction