missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrillin 1 Antibody (CL0225) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52885AF488
This item is not returnable.
View return policy
Description
Fibrillin 1 Monoclonal antibody specifically detects Fibrillin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Fibrillin 1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Asprosin, FBN, fibrillin 1, fibrillin 1 (Marfan syndrome), fibrillin 15, fibrillin-1, MASS, MFS1, OCTD, SGS, SSKS, WMS | |
| This antibody was developed using a recombinant protein derived from P35555, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Extracellular Matrix | |
| 2200 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0225 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction