missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrillarin Recombinant Rabbit Monoclonal Antibody (23GB5735), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552901
This item is not returnable.
View return policy
Description
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
Specifications
| Fibrillarin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| fbl | |
| 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14113, 2091, 292747 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5735 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22087, P22509, P35550 | |
| fbl | |
| A synthesized peptide derived from human Fibrillarin (270-321AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction