missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrillarin/FBLL1 Recombinant Rabbit Monoclonal Antibody (HL2588)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556708
This item is not returnable.
View return policy
Description
Fibrillarin/FBLL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Fibrillarin, encoded by the FBL gene (also known as FBLL1), is a highly conserved nucleolar protein that plays a critical role in the biogenesis of ribosomes. It is a component of the small nucleolar ribonucleoprotein (snoRNP) complexes and is primarily involved in the chemical modification of ribosomal RNA (rRNA), specifically in the methylation of the 2'-O-ribose residues of rRNA. This modification is essential for the proper processing and maturation of pre-rRNA into functional ribosomal subunits, which are crucial for protein synthesis within the cell. Fibrillarin is not only important for ribosome production but also for maintaining the structural integrity of the nucleolus, the site of ribosome assembly. Dysregulation or mutations in the FBL gene have been associated with various human diseases, including cancer, where altered ribosome biogenesis can contribute to uncontrolled cell proliferation. Additionally, fibrillarin has been studied in the context of autoimmune diseases, such as systemic sclerosis, where it can serve as an autoantigen. Understanding the function and regulation of fibrillarin is vital for insights into cellular growth, protein synthesis, and the pathogenesis of related diseases.
Specifications
| Fibrillarin/FBLL1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| A6NHQ2, P22087, P22509, P35550, Q80WS3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14113, 2091, 237730, 292747, 345630, 363563 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2588 | |
| Unconjugated | |
| FBLL1 | |
| fbl, FBLL1 | |
| Recombinant fragment of human fibrillarin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction