missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrillarin Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£173.00 - £406.00
Specifications
| Antigen | Fibrillarin |
|---|---|
| Dilution | Western Blot 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000 |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18091804
|
Novus Biologicals
NBP2-46881-0.025ML |
0.025 mL |
£173.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18072676
|
Novus Biologicals
NBP2-46881 |
0.1 mL |
£406.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibrillarin Polyclonal antibody specifically detects Fibrillarin in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
| Fibrillarin | |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Chicken | |
| Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers | |
| EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
| FBL | |
| IgY | |
| IgY purified | |
| 34.5 kDa |
| Western Blot 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| P22087 | |
| 2091 | |
| Full length human Fibrillarin expressed in and purified from E.Coli. [UniProt# P22087] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title