Learn More
FIBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15968-1-AP-20UL
 View more versions of this product
Product Code. 16875384
  • £135.00 / 20µL
Description
Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
FIBP
Polyclonal
Unconjugated
FIBP
FGFIBP, FIBP, FIBP 1
Rabbit
Antigen Affinity Chromatography
RUO
58249, 9158
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43427, Q9JI19
Fibp
FIBP Fusion Protein Ag8743
20 μL
Primary
Human, Mouse