missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15968-1-AP-150UL
This item is not returnable.
View return policy
Description
Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| FIBP | |
| Polyclonal | |
| Unconjugated | |
| FIBP | |
| FGFIBP, FIBP, FIBP 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58249, 9158 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43427, Q9JI19 | |
| Fibp | |
| FIBP Fusion Protein Ag8743 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction