Learn More
FIBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589306
Description
Immunogen sequence: MTSELDIFVG NTTLIDEDVY RLWLDGYSVT DAVALRVRSG ILEQTGATAA VLQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGIT LKSCRRQFDN FKRVFKVVEE MRGSLVDNIQ QHFLLSDRLA RDYAAIVFFA NNRFETGKKK LQYLSFGDFA FCAELMIQNW TLGAVDSQMD DMDMDLDKEF LQDLKELKVL VADKDLLDLH KSLVCTALRG KLGVFSEMEA NFKNLSRGLV NVAAKLTHNK DVRDLFVDLV EKFVEPCRSD HWPLSDVRFF LNQYSASVHS LDGFRHQALW DRYMGTLRGC LLRLYHD; Positive Samples: BXPC-3, Mouse skeletal muscle; Cellular Location: Endomembrane system, Nucleus, Peripheral membrane protein
Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| FIBP | |
| Polyclonal | |
| Unconjugated | |
| FIBP | |
| 2010004G08Rik, 2010005N21Rik, 3010027N18Rik, Acidic fibroblast growth factor intracellular-binding protein, aFGF intracellular binding protein, aFGF intracellular-binding protein, FGF intracellular binding protein, FGF1 intracellular binding protein, FGF-1 intracellular-binding protein, FGFIBP, Fibp, FIBP-1, fibroblast growth factor (acidic) intracellular binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 58249, 9158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
| O43427, Q9JI19 | |
| FIBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human FIBP (NP_0042052) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.