Learn More
Invitrogen™ FIBP Monoclonal Antibody (1E5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524425
Description
Peptide Sequence: MTSELDIFVG NTTLIDEDVY RLWLDGYSVT DAVALRVRSG ILEQTGATAA VLQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGIT LKSCRRQFDN FKRVFKVVEE MRGSLVDNIQ QHFLLSDRLA RDYAAIVFFA NNRFETGKKK LQYLSFGDFA FCAELMIQNW TLGAVDSQMD DMDMDLDKEF LQDLKELKVL VADKDLLDLH KSLVCTALRG KLGVFSEMEA NFKNLSRGLV NVAAKLTHNK DVGDLFVDLV EKFVEPCRSD HWPLSDVRFF LNQYSASVHS LDGFRHQALW DRYMGTLRGC LLRLYHD.
Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| FIBP | |
| Monoclonal | |
| Unconjugated | |
| FIBP | |
| 2010004G08Rik; 2010005N21Rik; 3010027N18Rik; Acidic fibroblast growth factor intracellular-binding protein; aFGF intracellular binding protein; aFGF intracellular-binding protein; FGF intracellular binding protein; FGF1 intracellular binding protein; FGF-1 intracellular-binding protein; FGFIBP; Fibp; FIBP-1; fibroblast growth factor (acidic) intracellular binding protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 9158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 1E5 | |
| PBS with no preservative; pH 7.4 | |
| O43427 | |
| FIBP | |
| FIBP (AAH14388, 1 a.a. ~ 357 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.