Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
£473.00 - £755.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11874511
|
Thermo Scientific™
00388 |
5mL Round-bottom Tube |
£550.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12796947
|
Thermo Scientific™
00456 |
7mL Round-bottom Tube |
£490.00
Each |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11844511
|
Thermo Scientific™
010-0138 |
50mL Round-bottom Tube (Set of 2) |
£473.00
Pack of 2 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11898362
|
Thermo Scientific™
75100136 |
50mL Conical Tube (Set of 2) |
£755.00
Pack of 2 |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11808372
|
Thermo Scientific™
75101073 |
15 mL Conical Tube (Set of 2) |
£755.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:- Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.