Learn More
FIBCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25125-1-AP-20UL
 View more versions of this product
Product Code. 16823675
  • £135.00 / 20µL
Description
FIBCD1 is a single-pass membrane protein. It contains 1 fibrinogen C-terminal domain. The exact function of FIBCD1 remains unknown.
Specifications
Specifications
FIBCD1
Polyclonal
Unconjugated
FIBCD1
FIBCD1
Rabbit
Antigen Affinity Chromatography
RUO
84929
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.73 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N539
FIBCD1
FIBCD1 Fusion Protein Ag18355
20 μL