missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FIBCD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15576072
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15576072 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576072 Supplier Invitrogen™ Supplier No. PA543502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: DGYPLTVADY SGTAGDSLLK HSGMRFTTKD RDSDHSENNC AAFYRGAWWY Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

FIBCD1 is a single-pass membrane protein. It contains 1 fibrinogen C-terminal domain. The exact function of FIBCD1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIBCD1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FIBCD1
Gene Accession No. A2AV25, Q8N539
Gene Alias AI448887; Fibcd1; fibrinogen C domain containing 1; fibrinogen C domain-containing protein 1; fibrinogen C domain-containing protein 1; uncharacterized protein LOC567525; LOW QUALITY PROTEIN: fibrinogen C domain-containing protein 1; RGD1309097; si:dkey-54o1.1; UNQ701/PRO1346
Gene Symbols FIBCD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FIBCD1 (aa 362-411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84929, 98970
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.