missing translation for 'onlineSavingsMsg'
Learn More

Fialuridine, TRC

Product Code. 30400029
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400029 5 mg -
30396605 100 mg 100mg
30402404 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400029 Supplier Toronto Research Chemicals Supplier No. TRCF3350005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69123-98-4
Molecular Formula C9 H10 F I N2 O5
Molecular Weight (g/mol) 372.09
Synonym 2,4(1H,3H)-Pyrimidinedione, 1-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodo-, 1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodo-2,4(1H,3H)-pyrimidinedione, 1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodouracil, 1-(2'-Deoxy-2'-fluoro-β-D-arabinofuranosyl)-5-iodouracil, 5-Iodo-2'-fluoroarauracil, FIAU, Fialuridine, Fluoroiodoarauracil, NSC 678514
IUPAC Name 1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@@H](F)[C@@H]1O)N2C=C(I)C(=O)NC2=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H10FIN2O5/c10-5-6(15)4(2-14)18-8(5)13-1-3(11)7(16)12-9(13)17/h1,4-6,8,14-15H,2H2,(H,12,16,17)/t4-,5+,6-,8-/m1/s1
Formula Weight 371.9618
Chemical Name or Material Fialuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.