missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress FHZ

Product Code. 16452648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16452648

Brand: MedChemExpress HYU00440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FHZ is a fluorescent probe.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Product Type Dye Reagents
Quantity 5 mg
Shipping Condition Room temperature
CAS 1883737-63-0
Molecular Weight (g/mol) 478.49 g/mol
Solubility DMSO : 100 mg/mL (208.99 mM; Need ultrasonic)
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.