missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116238
This item is not returnable.
View return policy
Description
FHOD1 Polyclonal Antibody for Western Blot
The function of this protein remains unknown.
Specifications
| FHOD1 | |
| Polyclonal | |
| Unconjugated | |
| FHOD1 | |
| BC017144; FH1/FH2 domain-containing protein 1; Fhod1; FHOS; FHOS1; formin homolog overexpressed in spleen 1; formin homology 2 domain containing 1; formin homology 2 domain-containing protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 234686, 29109 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6P9Q4, Q9Y613 | |
| FHOD1 | |
| KLH conjugated synthetic peptide between 985-1013 amino acids from the C-terminal region of human FHOD1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction