missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FHOD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28958
This item is not returnable.
View return policy
Description
FHOD1 Polyclonal antibody specifically detects FHOD1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| FHOD1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FH1/FH2 domain-containing protein 1, FHOS1, FHOSFormin homolog overexpressed in spleen 1, formin homology 2 domain containing 1, Formin homology 2 domain-containing protein 1 | |
| Between 1050 and 1100 | |
| 100 μg | |
| Apoptosis | |
| 29109 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction