missing translation for 'onlineSavingsMsg'
Learn More

FHOD1 Antibody, Novus Biologicals™

Product Code. 18378529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378529 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378529 Supplier Novus Biologicals Supplier No. H00029109B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

FHOD1 Polyclonal antibody specifically detects FHOD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHOD1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.4)
Gene Accession No. NP_037373.2
Gene Alias FH1/FH2 domain-containing protein 1, FHOS1, FHOSFormin homolog overexpressed in spleen 1, formin homology 2 domain containing 1, Formin homology 2 domain-containing protein 1
Host Species Mouse
Immunogen FHOD1 (NP_037373.2, 1 a.a. - 1164 a.a.) full-length human protein. MAGGEDRGDGEPVSVVTVRVQYLEDTDPFACANFPEPRRAPTCSLDGALPLGAQIPAVHRLLGAPLKLEDCALQVSPSGYYLDTELSLEEQREMLEGFYEEISKGRKPTLILRTQLSVRVNAILEKLYSSSGPELRRSLFSLKQIFQEDKDLVPEFVHSEGLSCLIRVGAAADHNYQSYILRALGQLMLFVDGMLGVVAHSDTIQWLYTLCASLSRLVVKTALKLLLVFVEYSENNAPLFIRAVNSVASTTGAPPWANLVSILEEKNGADPELLVYTVTLINKTLAALPDQDSFYDVTDALEQQGMEALVQRHLGTAGTDVDLRTQLVLYENALKLEDGDIEEAPGAGGRRERRKPSSEEGKRSRRSLEGGGCPARAPEPGPTGPASPVGPTSSTGPALLTGPASSPVGPPSGLQASVNLFPTISVAPSADTSSERSIYKARFLENVAAAETEKQVALAQGRAETLAGAMPNEAGGHPDARQLWDSPETAPAARTPQSPAPCVLLRAQRSLAPEPKEPLIPASPKAEPIWELPTRAPRLSIGDLDFSDLGEDEDQDMLNVESVEAGKDIPAPSPPLPLLSGVPPPPPLPPPPPIKGPFPPPPPLPLAAPLPHSVPDSSALPTKRKTVKLFWRELKLAGGHGVSASRFGPCATLWASLDPVSVDTARLEHLFESRAKEVLPSKKAGEGRRTMTTVLDPKRSNAINIGLTTLPPVHVIKAALLNFDEFAVSKDGIEKLLTMMPTEEERQKIEEAQLANPDIPLGPAENFLMTLASIGGLAARLQLWAFKLDYDSMEREIAEPLFDLKVGMEQLVQNATFRCILATLLAVGNFLNGSQSSGFELSYLEKVSEVKDTVRRQSLLHHLCSLVLQTRPESSDLYSEIPALTRCAKVDFEQLTENLGQLERRSRAAEESLRSLAKHELAPALRARLTHFLDQCARRVAMLRIVHRRVCNRFHAFLLYLGYTPQAAREVRIMQFCHTLREFALEYRTCRERVLQQQQKQATYRERNKTRGRMITETEKFSGVAGEAPSNPSVPVAVSSGPGRGDADSHASMKSLLTSRPEDTTHNRRSRGMVQSSSPIMPTVGPSTASPEEPPGSSLPSDTSDEIMDLLVQSVTKSSPRALAARERKRSRGNRKSLRRTLKSGLGDDLVQALGLSKGPGLEV
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 29109
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.