missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHOD1 Antibody (M352), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23340
This item is not returnable.
View return policy
Description
FHOD1 Monoclonal antibody specifically detects FHOD1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| FHOD1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| M352 | |
| Western Blot 1:250, Immunohistochemistry 1:50 | |
| FH1/FH2 domain-containing protein 1, FHOS1, FHOSFormin homolog overexpressed in spleen 1, formin homology 2 domain containing 1, Formin homology 2 domain-containing protein 1 | |
| FHOD1 recombinant protein that included the full length human FHOD1 sequence. This sequence has 50% homology to human FHOD3, and low homology to other formin family members. [UniProt# Q9Y613] | |
| 0.1 mL | |
| Apoptosis | |
| 29109 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction