missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550300
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total FHL5 protein.
The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| FHL5 | |
| Polyclonal | |
| Unconjugated | |
| FHL5 | |
| 1700027G07Rik; Act; activator of cAMP-responsive element modulator (CREM) in testis; activator of cAMP-responsive element modulator in testis; Activator of CREM in testis; dJ393D12.2; FHL5; FHL-5; four and a half LIM domains 5; four and a half LIM domains protein 5; LIM protein ACT; LIM-only protein ACT; RP3-393D12.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9457 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5TD97 | |
| FHL5 | |
| Fusion protein corresponding to a region derived from internal residues of human four and a half LIM domains 5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction