missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL5 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27588CL1
This item is not returnable.
View return policy
Description
FHL5 Polyclonal antibody specifically detects FHL5 in Human,Mouse samples. It is validated for Western Blot
Specifications
| FHL5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| activator of cAMP-responsive element modulator (CREM) in testis, Activator of cAMP-responsive element modulator in testis, Activator of CREM in testis, ACTKIAA0776, dJ393D12.2, FHL-5, FLJ33049, four and a half LIM domains 5,1700027G07Rik, four and a half LIM domains protein 5, LIM protein ACT, RP3-393D12.2 | |
| Synthetic peptide corresponding to the N-terminus of ACT protein | |
| 0.1 mL | |
| Signal Transduction | |
| 9457 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction