missing translation for 'onlineSavingsMsg'
Learn More

FHL5 Antibody, Biotin, Novus Biologicals™

Product Code. 18401872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401872 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401872 Supplier Novus Biologicals Supplier No. NBP227588B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FHL5 Polyclonal antibody specifically detects FHL5 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FHL5
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias activator of cAMP-responsive element modulator (CREM) in testis, Activator of cAMP-responsive element modulator in testis, Activator of CREM in testis, ACTKIAA0776, dJ393D12.2, FHL-5, FLJ33049, four and a half LIM domains 5,1700027G07Rik, four and a half LIM domains protein 5, LIM protein ACT, RP3-393D12.2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of ACT protein
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9457
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.