missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112912
This item is not returnable.
View return policy
Description
FHL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.
Specifications
| FHL3 | |
| Polyclonal | |
| Unconjugated | |
| FHL3 | |
| FHL3; FHL-3; four and a half LIM domains 3; four and a half LIM domains protein 3; LIM-only protein FHL3; MGC23614; MGC8696; skeletal muscle LIM-protein 2; SLIM2; SLIM-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2275 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13643 | |
| FHL3 | |
| Recombinant Human Four and a half LIM domains protein 3 protein (1-280AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction