missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHL2 Recombinant Rabbit Monoclonal Antibody (23GB5860), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FHL2 |
|---|---|
| Clone | 23GB5860 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088545
![]() |
Invitrogen™
MA552934 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089493
|
Invitrogen™
MA552935 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FHL2 Recombinant Monoclonal antibody specifically detects FHL2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FHL2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O35115, O70433, Q14192 | |
| 14200, 2274, 63839 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5860 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AAG11; aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; DRAL; Fhl2; FHL-2; four and a half LIM domains 2; four and a half LIM domains protein 2; LIM domain protein DRAL; OTTHUMP00000203164; OTTHUMP00000203168; OTTHUMP00000203169; skeletal muscle LIM-protein 3; SLIM 3; SLIM3; SLIM-3 | |
| A synthesized peptide derived from human FHL2 (20-279AA). | |
| Primary | |
| -20°C | |
| Fhl2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title