missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21619-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. This protein is thought to have a role in the assembly of extracellular membranes. Also, this gene is down-regulated during transformation of normal myoblasts to rhabdomyosarcoma cells and the encoded protein may function as a link between presenilin-2 and an intracellular signaling pathway. Multiple alternatively spliced variants encoding different isoforms have been identified.Specifications
| FHL2 | |
| Polyclonal | |
| Unconjugated | |
| FHL2 | |
| AAG11, DRAL, FHL 2, FHL2, four and a half LIM domains 2, LIM domain protein DRAL, Skeletal muscle LIM protein 3, SLIM 3, SLIM3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14200, 2274, 63839 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35115, O70433, Q14192 | |
| Fhl2 | |
| FHL2 Fusion Protein Ag1378 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction