missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL2 Antibody (AT21D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61162CL1
This item is not returnable.
View return policy
Description
FHL2 Monoclonal antibody specifically detects FHL2 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| FHL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11 | |
| Recombinant human FHL2 (1-279aa) purified from E. coli. | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 2274 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| AT21D11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction