Learn More
Invitrogen™ FHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596331
Description
Immunogen sequence: RDPLQGKKYV QKDGHHCCLK CFDKFCANTC VECRKPIGAD SKEVHYKNRF WHDTCFRCAK CLHPLANETF VAKDNKILCN KCTTREDSPK CKGCFKAIVA GDQNVEYKGT VWHKDCFTCS NCKQVIGTGS FFPKGEDFYC VTCHETKFAK HCVKCNKAIT SGGITYQDQP WHADCFVCVT CSKKLAGQRF TAVEDQYYCV DCYKNFVAKK CAGCKNPITG FGKGSSVVAY EGQSWHDYCF HCKKCSVNLA NKRFVFHQEQ VYCPDCAKKL; Positive Samples: U-251MG, 22Rv1, MCF7, Mouse kidney; Cellular Location: Cytoplasm, Nucleus, cytosol.
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.
Specifications
| FHL1 | |
| Polyclonal | |
| Unconjugated | |
| FHL1 | |
| bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1a; four and a half LIM domains 1b; four and a half LIM domains 1b; four and a half LIM domains protein 1b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1a; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBMX1A; RBMX1B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14199, 2273 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97447, Q13642 | |
| FHL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-296 of human FHL1 (NP_0011531711). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.