missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHIT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 719000
This item is not returnable.
View return policy
Description
FHIT Polyclonal Antibody for Western Blot, IHC, ICC/IF, ELISA
Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.
Specifications
| FHIT | |
| Polyclonal | |
| Unconjugated | |
| FHIT | |
| AP3A hydrolase; AP3Aase; AW045638; bis(5'-adenosyl)-triphosphatase; bis5'-adenosyl-triphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; dinucleosidetriphosphatase; FHIT; Fra14A2; FRA3B; fragile histidine triad; fragile histidine triad gene; fragile histidine triad protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2272, 60398 | |
| -20°C | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P49789, Q9JIX3 | |
| FHIT | |
| Full length human FHIT fusion protein with GST (glutathione amino transferase). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction