missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHIT Mouse anti-Human, Clone: 4D6G1, Proteintech
Mouse Monoclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | FHIT |
|---|---|
| Clone | 4D6G1 |
| Concentration | 0.67 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
Description
Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.Specifications
| FHIT | |
| 0.67 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AP3A hydrolase, AP3Aase, Bis(5 adenosyl) triphosphatase, Dinucleosidetriphosphatase, FHIT, FRA3B, fragile histidine triad gene | |
| FHIT | |
| IgG1 | |
| Protein A | |
| Antibody |
| 4D6G1 | |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P49789 | |
| 2272 | |
| FHIT Fusion Protein Ag14471 | |
| Primary | |
| -20°C | |
| FHIT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title