missing translation for 'onlineSavingsMsg'
Learn More

FH1, MedChemExpress

Product Code. 16481347
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481347 50 mg 50mg
16415118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481347 Supplier Medchem Express Supplier No. HY12346
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FH1 (NSC 12407) enhances hepatocyte functions, and promotes the differentiation of induced pluripotent stem (iPS)-derived hepatocytes toward a phenotype more mature and the maturation of well-differentiated cultures of hepatocyte-like cells (iHeps).

FH1 (NSC 12407) enhances hepatocyte functions, and promotes the differentiation of induced pluripotent stem (iPS)-derived hepatocytes toward a phenotype more mature and the maturation of well-differentiated cultures of hepatocyte-like cells (iHeps).

Specifications

Chemical Name or Material FH1
CAS 2719-05-3
Color Orange
Purity Grade Notes Research
Molecular Formula C17H18N2O2
Quantity 50 mg
Synonym NSC 12407 BRD-K4477
Solubility Information DMSO : 13 mg/mL (46.04 mM; Need ultrasonic and warming)
SMILES CC(NC1=CC=C(CC2=CC=C(NC(C)=O)C=C2)C=C1)=O
Molecular Weight (g/mol) 282.34
Formula Weight 282.34
Percent Purity 99.42%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.