Learn More
Invitrogen™ FGR Monoclonal Antibody (6G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515490
Description
MA5-15490 targets FGR in indirect ELISA, IHC and WB applications and shows reactivity with Human and mouse samples. The MA5-15490 immunogen is purified recombinant fragment of human FGR expressed in E. Coli. MA5-15490 detects FGR which has a predicted molecular weight of approximately 56kDa.
FGR is a SRC-related kinase present in hematopoietic cells. FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors. This protein contains N-terminal sites for myristylation and palmitoylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. It localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this protein.
Specifications
| FGR | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| P09769, P14234 | |
| FGR | |
| Purified recombinant fragment of human FGR expressed in E. Coli. | |
| RUO | |
| 14191, 2268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6G2 | |
| Unconjugated | |
| FGR | |
| c-fgr; c-fgr protooncogene; c-src2; c-src-2 proto-oncogene; EC 2.7.10.2; feline Gardner-Rasheed sarcoma viral oncogene; feline Gardner-Rasheed sarcoma viral oncogene homolog; FGR; FGR protein; FGR proto-oncogene, Src family tyrosine kinase; FLJ43153; Gardner-Rasheed feline sarcoma viral (Fgr) oncogene homolog; Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog; MGC75096; OTTHUMP00000003603; OTTHUMP00000003604; p55c-fgr; p55-c-fgr protein; p55-Fgr; p58c-Fgr; p58-Fgr; proto-oncogene c-Fgr; proto-oncogene tyrosine-protein kinase FGR; SRC2; Tyrosine-protein kinase Fgr; v-fgr feline Gardner-Rasheed sarcoma viral oncogene homolog | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.