Learn More
FGL2 Mouse anti-Human, Unconjugated, Clone: 2C9C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67152-1-IG-150UL
This item is not returnable.
View return policy
Description
The FGL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FGL2. This antibody recognizes human antigen. The FGL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FGL2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 2C9C5 | |
| Unconjugated | |
| 150 μL | |
| 10875 | |
| IgG2b |