missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FGL2/Fibroleukin Monoclonal antibody specifically detects FGL2/Fibroleukin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay
Specifications
Specifications
| Antigen | FGL2/Fibroleukin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay |
| Classification | Monoclonal |
| Clone | 6D9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.5μg/mL, Functional |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_006673 |
| Gene Alias | fibrinogen-like 2, Fibrinogen-like protein 2, fibroleukin, pT49T49 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?