Learn More
Invitrogen™ FGL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109725
Description
Immunogen sequence: MAKVFSFILV TTALTMGREI SALEDCAQEQ MRLRAQVRLL ETRVKQQQVK IKQLLQENEV QFLDKGDENT VIDLGSKRQY ADCSEIFNDG YKLSGFYKIK PLQSPAEFSV YCDMSDGGGW TVIQRRSDGS ENFNRGWKDY ENGFGNFVQK HGEYWLGNKN LHFLTTQEDY TLKIDLADFE KNSRYAQYKN FKVGDEKNFY ELNIGEYSGT AGDSLAGNFH PEVQWWASHQ RMKFSTWDRD HDNYEGNCAE EDQSGWWFNR CHSANLNGVY YSGPYTAKTD NGIVWYTWHG WWYSLKSVVM KIRPNDFIPN VI.
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
Specifications
| FGL1 | |
| Polyclonal | |
| Unconjugated | |
| FGL1 | |
| Fgl1; fibrinogen like 1; fibrinogen-like 1; fibrinogen-like protein 1; Fibrinogen-related protein 1; fibronigen-like protein 1; Frep1; Hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; HFREP1; HFREP-1; HP-041; HPS; Lfire1; LFIRE-1; Liver fibrinogen-related protein 1; Liver fibrinogen-related protein-1; Mfire1; Mfire-1; Mfrep-1; MGC12455; OTTHUMP00000122468 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2267 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q08830 | |
| FGL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-312 of human FGL1 (NP_004458.3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.