missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
£155.00 - £430.00
Specifications
| Antigen | FGL1 |
|---|---|
| Concentration | 0.71 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17780524
![]() |
Invitrogen™
PA5119116 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17713394
|
Invitrogen™
PA5121993 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGL1 Polyclonal Antibody for Western Blot, IHC (P)
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.Specifications
| FGL1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| FGL1 | |
| Fgl1; fibrinogen like 1; fibrinogen-like 1; fibrinogen-like protein 1; Fibrinogen-related protein 1; fibronigen-like protein 1; Frep1; Hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; HFREP1; HFREP-1; HP-041; HPS; Lfire1; LFIRE-1; Liver fibrinogen-related protein 1; Liver fibrinogen-related protein-1; Mfire1; Mfire-1; Mfrep-1; MGC12455; OTTHUMP00000122468 | |
| FGL1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.71 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q08830 | |
| 2267 | |
| Recombinant protein Fibrinogen Like Protein 1. The antigen corresponds to amino acid range 23-312 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title