missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGIN-1-27, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Binds specifically and with high affinity to the peptide DBI receptor (benzodiazepine receptor) on mitochondrial membranes, inducing production of neurosteroids, which modulate GABA receptors (EC50 = 3 nM for increase in production of pregnenolone in glial cells). Thus it reduces anxiety without causing sedation.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 436.61 |
| Percent Purity | >98% |
| Chemical Name or Material | FGIN-1-27 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction