missing translation for 'onlineSavingsMsg'
Learn More

FGH10019, MedChemExpress

Product Code. 16459547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459547 5 mg 5mg
16433057 50 mg 50mg
16407008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16459547 Supplier Medchem Express Supplier No. HY16207
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FGH10019 is a novel sterol regulatory element-binding protein (SREBP) inhibitor with IC50 of 1 μM.

FGH10019 is a novel sterol regulatory element-binding protein (SREBP) inhibitor with IC50 of 1 μM.

Specifications

Chemical Name or Material FGH10019
CAS 1046045-61-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C18H19N3O2S2
Quantity 5 mg
Solubility Information DMSO : ≥ 38 mg/mL (101.74 mM)
SMILES CS(=O)(NC1=CC=C(C2=CSC(C3=CC(CCC)=NC=C3)=N2)C=C1)=O
Molecular Weight (g/mol) 373.49
Formula Weight 373.49
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.