missing translation for 'onlineSavingsMsg'
Learn More

FGGY carbohydrate kinase domain containing Antibody (3B9), Novus Biologicals™

Product Code. 18374369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374369 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374369 Supplier Novus Biologicals Supplier No. H00055277M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGGY carbohydrate kinase domain containing Monoclonal antibody specifically detects FGGY carbohydrate kinase domain containing in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGGY carbohydrate kinase domain containing
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3B9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060761
Gene Alias EC 2.7.1, EC 2.7.1.-, FGGY carbohydrate kinase domain containing, FGGY carbohydrate kinase domain-containing protein, FLJ10986
Host Species Mouse
Immunogen FLJ10986 (NP_060761, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MWLDHRAVSQVNRINETKHSVLQYVGGVMSVEMQAPKLLWLKENLREICWDKAGHFFDLPDFLSWKATGVTARSLCSLVCKWTYSAEKGWDDSFWKMIG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55277
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.