missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR5/FGFRL1 Antibody (RM0232-7L43), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12294R
This item is not returnable.
View return policy
Description
FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.
Specifications
| FGFR5/FGFRL1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGFRL1 | |
| Mouse recombinant FGFR5/FGFRL1 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0232-7L43 | |
| Western Blot | |
| FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
| Rat | |
| Protein A or G purified | |
| Angiogenesis | |
| 53834 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction