missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR5/FGFRL1 Antibody (004), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90466DL594
This item is not returnable.
View return policy
Description
FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for ELISA.
Specifications
| FGFR5/FGFRL1 | |
| Monoclonal | |
| DyLight 594 | |
| FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGFR5/FGFRL1 (NP_473412.1; Met 1-Pro 374). | |
| 0.1 mL | |
| Angiogenesis | |
| 53834 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 4 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction